Skip to Content
Merck
All Photos(1)

Key Documents

CLS3701

Corning® 384 well microplate

Tissue Culture (TC)-treated surface, clear polystyrene, flat bottom, sterile, lid, case of 100

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
clear polystyrene
flat bottom
size:
384 wells

material

clear bottom
clear polystyrene
flat bottom

sterility

sterile

feature

lid
skirt

packaging

case of 100

manufacturer/tradename

Corning 3701

size

384 wells

well volume

112 μL

well working volume

20-80 μL

wells

384 wells

color

clear

suitability

suitable for tissue

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384 well microplate is an ELISA microplate and has a tissue culture treated surface for optimal cell attachment.
  • Applicable for colorimetric assays
  • Sterilized by gamma radiation and nonpyrogenic
  • Individual alphanumeric codes enable well identification

Application

For colorimetric assays

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service