Skip to Content
Merck
All Photos(1)

Key Documents

TGP 55

95062833, mouse pancreas, Polymorph

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
mouse pancreas
growth mode:
Adherent
karyotype:
Not specified
morphology:
Polymorph
products:
Somatostatin and Insulin
receptors:
Not specified
relevant disease(s):
cancer

Product Name

TGP 55, 95062833

biological source

mouse pancreas

growth mode

Adherent

karyotype

Not specified

morphology

Polymorph

products

Somatostatin and Insulin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse pancreatic small cell anaplastic carcinoma

Cell Line Description

TGP 55 has been obtained from a small cell anaplastic carcinoma arising in the pancreas of a transgenic Tg(Ela-1SV40E)Bri 18 mouse these mice carry a pseudogene construct composed of a pancreatic-specific elastase-1 promoter linked to the coding region for SV40 T antigen and thereby represent an animal model for pancreatic carcinogenesis. TGP 55 cells are SV40T antigen positive and secrete somatostatin, it is one of a series of cell lines isolated from pancreatic tumours deposited at ECACC. Cells secrete somatostatin and insulin in culture. Adherent clumps of small cells.

Application

Study of exocrine pancreas carcinomas, hormone secretion studies.

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service