Skip to Content
Merck
All Photos(1)

Key Documents

NP cl.120

Reactivity: Anti-Measles Virus Nucleoprotein, 95040312

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0 Ag-14

Reactivity

Anti-Measles Virus Nucleoprotein

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

NP cl.120 monoclonal antibody is one of a range of well characterised measles virus antibodies used to measure synthesis and topological accumulation of measles virus antigen during the lytic cycle. This anti-nucleoprotein antibody is negative for haemagglutinin and hemolysin inhibition as well as neutralisation assays and gives immunofluorescence only on fixed tissue.

Immunogen

Measles virus (strain Hallé)

Application

Diagnosis, study of measles virus infection

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 1-5x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service