JEG3
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92120308, human placenta, Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human placenta
packaging
tube of 5 μg 92120308-DNA-5UG
pkg of vial of cells 92120308-1VL
growth mode
Adherent
karyotype
modal no. 71
morphology
Epithelial
products
Human chorionic gonadotrophin (hCG), somatomammotrophin
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human choriocarcinoma
Cell Line Description
One of 6 clones derived from cells implanted into a hamster cheek pouch and then propagated on irradiated feeder layers of human fibroblasts. Cell line releases chorionic gonadotrophin and somatomammotrophin and progesterone. It is also able to transform steroid precursors to oestrone and oestradiol. JEG3 has the same DNA profile as BeWo (Sigma Catalogue number 86082803); it was established by serial cloning of BeWo .
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin, 5% CO2; 37°C. Vacuolisation will occur at confluency.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service