Skip to Content
Merck
All Photos(1)

Key Documents

SW 626

91091203, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SW 626, 91091203

biological source

human colon

packaging

tube of 5 μg 91091203-DNA-5UG
pkg of vial of cells 91091203-1VL

growth mode

Adherent

karyotype

Hypertetraploid; modal no. 104

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian ovarian metastasis of a primary colon adenocarcinoma

Cell Line Description

Although originally thought to be of ovarian origin, a recent report has suggested that the SW 686 cell line might have been derived from an ovarian metastasis of a primary adenocarcinoma of the colon Furlong, et al., 1999. The SW 626 cell line was initiated by A. Leibovitz in January 1974 at the Scott and White Clinic, Temple, Texas from a surgical specimen from a cystadenocarcinoma of the ovary in a 46 year old female Caucasian. The histopathology of the specimen was determined to be grade III adenocarcinoma.

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; CO2 is not needed. Medium change after 4-5 days. 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service