Recommended Products
Product Name
GS-109-V-63, 90110503
biological source
human skin
growth mode
Adherent
karyotype
Modal no. 46
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian skin fibroblast Gardner′s syndrome
Cell Line Description
Derived from a 13 year old Caucasian male with Gardner′s syndrome, an autosomal dominant condition which predisposes to carcinoma and multiple polyps of the colon. The modal chromosome number was 46, occurring in 60% of cells.
Application
Tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11
D13S317: 11,12
D16S539: 12,13
D5S818: 11,12
D7S820: 9,10
THO1: 7,8
TPOX: 8,11
vWA: 14,17
CSF1PO: 11
D13S317: 11,12
D16S539: 12,13
D5S818: 11,12
D7S820: 9,10
THO1: 7,8
TPOX: 8,11
vWA: 14,17
Culture Medium
EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).
Subculture Routine
Split semi-confluent cultures 1:2 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Monolayer should form in a 25cm2 flask in 3-4 days.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service