MNNG/HOS (TE85, Clone F-5)
87070201, human bone, Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
MNNG/HOS (TE85, Clone F-5), 87070201
biological source
human bone
packaging
tube of 5 μg 87070201-DNA-5UG
pkg of vial of cells 87070201-1VL
growth mode
Adherent
karyotype
Modal no. 70, 71; hypertriploid
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian osteosarcoma, chemically transformed
Cell Line Description
A chemically transformed cell line (MNNG 0.01ug/ml) derived from an osteosarcoma of a 13 year female Caucasian. Grows to high saturation density and produces tumours in nude mice.
Application
Transformation and tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 8,11
vWA: 18
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 8,11
vWA: 18
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service