Skip to Content
Merck
All Photos(1)

Key Documents

HFFF2

86031405, human prostate (fetal), Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HFFF2, 86031405

biological source

human prostate (fetal)

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian foetal foreskin fibroblast

Cell Line Description

Derived from a 14-18 week old human foetus. The cells support growth of CMV and HSV. This cell line has a finite life span.

Application

Virus studies: CMV and HSV

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 11,12
D16S539: 11,13
D5S818: 11
D7S820: 8,12
THO1: 9.3
TPOX: 8,11
vWA: 16,17

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service