Skip to Content
Merck
All Photos(1)

Key Documents

B12

85042303, rat nerve, Glial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

B12, 85042303

biological source

rat nerve

packaging

tube of 5 μg 85042303-DNA-5UG
pkg of vial of cells 85042303-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Glial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat nervous tissue glial

Cell Line Description

Ethyl nitrosourea induced tumour cell line. Glial cells are useful in the research of the distribution of neurotransmitter synthesis and brain specific antigens among nerve and glia.

Application

Neurobiological studies

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service