Skip to Content
Merck
All Photos(1)

Key Documents

1.1E7

10070101, human pancreas (islets), Epitheloid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human pancreas (islets)
growth mode:
Adherent
karyotype:
Modal chromosome number 67-71
morphology:
Epitheloid
products:
1.1E7 cells have been shown to express insulin, glucokinase and IAPP by immunocytochemistry
receptors:
Not specified

Product Name

1.1E7, from human pancreas(islets), 10070101

biological source

human pancreas (islets)

growth mode

Adherent

karyotype

Modal chromosome number 67-71

morphology

Epitheloid

products

1.1E7 cells have been shown to express insulin, glucokinase and IAPP by immunocytochemistry

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Description

1.1E7 is a hybrid cell line formed by the electrofusion of a primary culture of human pancreatic islets with PANC-1, a human pancreatic ductal carcinoma cell line (ECACC catalogue number 87092802). 1.1E7 has been shown to be tumourigenic when transplanted into a SCID mouse host. The cell line has applications in the study of pancreatic cell biology. 1.1E7 cells provide a method of producing pure insulin secreting cells when stimulated . This is an alternative to the use of primary tissue in cell transplantation therapies for type 1 diabetes. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,12
D13S317: 11
D16S539: 11
D5S818: 11,13
D7S820: 8
THO1: 7,8
TPOX: 8,11
vWA: 15

Culture Medium

RPMI-1640 + 2 mM Glutamine + 10% FCS / FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 , i.e., seeding at 2-4 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 23hrs. At confluence 105 cells/cm2 can be expected.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Legal Information

Flatt P & McClenaghan N (2004) Insulin producing cell-line US patent number 20040005702 (A1). Also WO0210346 (A2).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service