Skip to Content
Merck
All Photos(1)

Key Documents

VP267

5092805, human breast, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

VP267, 05092805

biological source

human breast

packaging

tube of 5 μg 05092805-DNA-5UG
pkg of vial of cells 05092805-1VL

growth mode

Adherent

karyotype

Mean no. 82, modal no. 62, range 45-148

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Breast Cancer

Cell Line Description

Derived from a breast cancer biopsy removed from a 48 year-old female. The histology of the breast cancer biopsy was defined as Ductal Grade III, tumour cells did not stain positive for oestrogen receptors. The VP267 cell line was derived from the same individual as the cell line VP229. VP229 was derived from the primary tumour and VP267 was derived 14 months later from a local recurrence of the tumour. The individual was recorded to live for 1year after surgery performed to remove the tumour. The patient had received tamoxifen treatment. This cell line grows slowly and is relatively difficult to culture; please read the sub-culture routine information closely.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11,13
D16S539: 11,12
D5S818: 11
D7S820: 10,12
THO1: 7
TPOX: 8
vWA: 17

Culture Medium

MEBM (Clonetics, CC-3151) + MEGM SingleQuots (Clonetics, CC-4136) + 2mM Glutamine + 2% Foetal Bovine Serum (FBS). Fresh EGF (final concentration of 10ng/ml) should be added to the medium at each subculture.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. The VP series of breast cancer cell lines are very slow growing and relatively difficult to grow. Fresh EGF (final concentration 10ng/ml) s

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service