All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H13NO4
CAS Number:
Molecular Weight:
235.24
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
form
powder
storage temp.
−20°C
SMILES string
OC(=O)C1(CC1)NC(=O)OCc2ccccc2
InChI
1S/C12H13NO4/c14-10(15)12(6-7-12)13-11(16)17-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,13,16)(H,14,15)
InChI key
KHINKCGJKZSHAJ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service