C-28021
Hematopoietic Progenitor Expansion Medium DXF
Ready-to-use, 500 ml
Synonym(s):
Stem Cell Growth Medium
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Quality
Preparation Note
Storage and Stability
Subculture Routine
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
CD34+ Progenitor Cells are suitable for diverse studies, as well as for the directed differentiation into more committed blood cell phenotypes and endothelial lineages.
Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.
Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.
Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.
Protocols
In vitro culture of human hematopoietic stem cells in PromoCell’s serum-free/xeno-free HPC Expansion Medium DXF. Explore over 350 PromoCell products
Hematopoietic stem cell culture protocols for isolation, expansion, and differentiation of CD34+ cells including CFU assays.
Hematopoietic stem cell culture protocols for isolation, expansion, and differentiation of CD34+ cells including CFU assays.
Hematopoietic stem cell culture protocols for isolation, expansion, and differentiation of CD34+ cells including CFU assays.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service