AXYPCR02NC
PCR tubes with flat caps
no caps, thin wall, size 0.2 mL, clear, pkg of 10x1000tubes/cs
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Axygen Inc PCR-02-NC
Corning
packaging
pkg of 10x1000tubes/cs
size
0.2 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service