Skip to Content
Merck
All Photos(2)

Key Documents

AV46329

Sigma-Aldrich

Anti-XTP3TPA (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CDA03, Anti-MGC5627, Anti-RS21C6, Anti-XTP3-transactivated protein A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

19 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XTP3TPA(79077)

Immunogen

Synthetic peptide directed towards the middle region of human XTP3TPA

Application

Anti-XTP3TPA (AB1) antibody produced in rabbit has been used for western blotting at a concentration of 0.5μg/ml. It has also been used for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

XTP3TPA (XTP3-transactivated protein A) also referred as DCTPP1 or RS21C6 is a 170 amino acid protein expressed in embryonic and highly proliferating cells primarily in liver, kidney, ovary and testis with particularly high expression in cancer cells.DCTPP1 hydrolyses 5-formyl-dCTP and plays a crucial role in the balance of dCTP as well as facilitates the metabolism of deoxycytidine analogs; hence contribute to the preservation of genome integrity.

Sequence

Synthetic peptide located within the following region: KMDINRRRYPAHLARSSSRKYTELPHGAISEDQAVGPADIPCDSTGQTST

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beili Wu et al.
Journal of molecular biology, 367(5), 1405-1412 (2007-02-27)
RS21-C6, which is highly expressed in all vertebrate genomes and green plants, is proposed to have nucleoside triphosphate pyrophosphohydrolase activity. Here, we report the crystal structures of the core fragment of RS21-C6, named RSCUT, and the complex with the substrate
Cristina E Requena et al.
The Biochemical journal, 459(1), 171-180 (2014-01-29)
The size and composition of dNTP (deoxyribonucleoside triphosphate) pools influence the accuracy of DNA synthesis and consequently the genetic stability of nuclear and mitochondrial genomes. In order to keep the dNTP pool in balance, the synthesis and degradation of DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service