Skip to Content
Merck
All Photos(2)

Key Documents

AV39936

Sigma-Aldrich

Anti-HIF3A antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Hypoxia inducible factor 3, α subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIF3A(64344)

Immunogen

Synthetic peptide directed towards the C terminal region of human HIF3A

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

HIF3A is the alpha-3 subunit of one of several alpha/beta-subunit heterodimeric transcription factors that regulate many adaptive responses to low oxygen tension (hypoxia). The alpha-3 subunit lacks the transactivation domain found in factors containing either the alpha-1 or alpha-2 subunits. It is thought that factors containing the alpha-3 subunit are negative regulators of hypoxia-inducible gene expression.The protein encoded by this gene is the alpha-3 subunit of one of several alpha/beta-subunit heterodimeric transcription factors that regulate many adaptive responses to low oxygen tension (hypoxia). The alpha-3 subunit lacks the transactivation domain found in factors containing either the alpha-1 or alpha-2 subunits. It is thought that factors containing the alpha-3 subunit are negative regulators of hypoxia-inducible gene expression. At least three transcript variants encoding three different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: PGSLQDPSTPLLNLNEPLGLGPSLLSPYSDEDTTQPGGPFQPRAGSAQAD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Janaszak-Jasiecka et al.
Scientific reports, 6, 22775-22775 (2016-03-10)
Hypoxia-inducible factors (HIF) are heterodimeric transcription factors that allow cells to adapt and survive during hypoxia. Regulation of HIF1A and HIF2A mRNA is well characterized, whereas HIF3A mRNA regulation and function are less clear. Using RNA-Seq analysis of primary human
Ming-Ching Lee et al.
Lung cancer (Amsterdam, Netherlands), 139, 157-164 (2019-12-07)
Thymoma is a rare epithelial tumor arising from the thymus in the anterior mediastinum. Nearly 50% of patients with thymoma develop myasthenia gravis, which is an indication of a poor long-term prognosis. Here, we identified specific and effective molecular markers
Kai Wu et al.
IUBMB life, 69(9), 660-667 (2017-07-08)
Asiatic acid (AA) could attenuate ischemia/reperfusion induced myocardial apoptosis through upregulating the Akt/GSK-3β/HIF-1α pathway. HIF-3α is a negative regulator of HIF-1α, whose mRNA is a potential target of miR-1290. AA could upregulate miR-1290 in non-small-cell lung cancer A549 cells. This
Maciej Jaskiewicz et al.
EXCLI journal, 21, 454-469 (2022-04-09)
The adaptive response to hypoxia involves the transcriptional induction of three transcription factors called hypoxia inducible factor alpha 1, 2 and 3 (HIF-1α, HIF-2α, and HIF-3α) which dimerize with constitutively expressed beta chains that together form the HIF-1, -2 and
Marcin Serocki et al.
Angiogenesis, 21(2), 183-202 (2018-02-01)
The decline of oxygen tension in the tissues below the physiological demand leads to the hypoxic adaptive response. This physiological consequence enables cells to recover from this cellular insult. Understanding the cellular pathways that mediate recovery from hypoxia is therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service