Skip to Content
Merck
All Photos(1)

Key Documents

AL11430

Sigma-Aldrich

DMT-locA(bz) Phosphoramidite

configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H52N7O8P
CAS Number:
Molecular Weight:
885.94
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

biological source

non-animal source (BSE/TSE no Risk)

Quality Level

product line

Proligo Reagents

Assay

≥98.0% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

impurities

≤3 wt. % Residual Solvent Content
<0.4% Water Content (Karl Fischer)
<0.5% Single unspecified Impurity (reversed phase HPLC)

color

white to off-white

solubility

soluble, clear, colorless

absorption

<0.1 in acetonitrile at 0.2 M

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for ABI

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

DMT-locA(bz) Phosphoramidite belongs to the group of LNA phosphoramidite. It is known as DMT-locAdenosine(N6-Benzoyl)- ß-Cyanoethylphosphoramidite. The synthesis cycle for Locked Nucleic Acid (LNA) oligonucleotides consists of the same series of reactions as the cycle that is employed for DNA monomers. However, the rate of coupling for LNA monomers is slower compared to that of DNA monomers (a coupling time of 8 minutes is recommended for LNA monomers compared to 90 seconds for DNA monomers).

With the exception of the LNA monomers, LNA synthesis is accomplished with the same reagents as DNA synthesis. LNA phosphoramidites from Merck are diluted with dry acetonitrile, except for locMeC(bz)-phosphoramidite. This phosphoramidite requires the application of a cosolvent to prevent crystallization from the solution on the synthesizer. Dichloromethane or tetrahydrofuran (THF) can be applied as co-solvents with acetonitrile to completely dissolve locMeC(bz)-phosphoramidite.

Features and Benefits

Key Features of LNA Phosphoramidites :

  • LNA oligonucleotides are prepared by phosphoramidite chemistry
  • Standard DNA synthesizer platforms can be employed. No change is required in the reagents commonly used for DNA synthesis
  • To further enhance the hybridization characteristics of LNA, 5-methyl-cytidine is employed instead of cytidine
  • LNA monomers are as soluble in acetonitrile as their DNA counterparts (except for the 5-methyl-cytidine derivative, which requires the application of 10-20%, dichloromethane as a co-solvent)
  • Mixmer oligonucleotides containing LNA, DNA and/or RNA monomers can be assembled easily
  • LNA oligonucleotides with predefined melting temperatures (Tm) can be designed and prepared

Other Notes

LNA phosphoramidites from Merck are diluted with dry acetonitrile, except for locMeC(bz)-phosphoramidite. This phosphoramidite requires the application of a cosolvent to prevent crystallization from the solution on the synthesizer. Dichloromethane or tetrahydrofuran(THF) can be applied as co-solvents with acetonitrile to completely dissolve locMeC(bz)-phosphoramidite.

Legal Information

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service