Skip to Content
Merck
All Photos(2)

Key Documents

A8376

Sigma-Aldrich

Acylase I from porcine kidney

Grade II, salt-free, lyophilized powder, 300-1,500 units/mg protein

Synonym(s):

Aminoacylase, N-Acylamino acid amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Grade II

Quality Level

form

salt-free, lyophilized powder

specific activity

300-1,500 units/mg protein

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Acylase I from porcine kidney has been used to study the acylase I-catalyzed deacetylation of various S-alkyl-N-acetyl-L-cysteines and their carbon and oxygen analogues . Acylase I may be useful to catalyze N-acetyl amino acids to enantiomerically pure L-amino acids .

Biochem/physiol Actions

Acylase I is a zinc metalloprotein that catalyzes the kinetic resolution of unnatural and rarely occurring α-amino acids. Its enantioselectivity for the hydrolysis of N-acyl L-α-amino acids is nearly absolute, yet it accepts substrates having a wide range of structure and functionality. Acylase I catalyzes the deacetylation of N-acetyl-L-cysteine and S-alkyl-N-acetyl-L-cysteines. n-Butylmalonic acid is an inhibitor of acylase I. S-alkyl-N-acetyl-L-cysteines with short (C0-C3) and unbranched S-alkyl substituents have been found to be good acylase I substrates.

Unit Definition

One unit will hydrolyze 1.0 μmole of N-acetyl-L-methionine per hr at pH 7.0 at 25 °C.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic resolution of unnatural and rarely occurring amino acids: enantioselective hydrolysis of N-acyl amino acids catalyzed by acylase I
Chenault HK, et al.
Journal of the American Chemical Society, 111(16), 6354-6364 (1989)
V Uttamsingh et al.
Chemical research in toxicology, 11(7), 800-809 (1998-07-22)
The aminoacylase that catalyzes the hydrolysis of N-acetyl-L-cysteine (NAC) was identified as acylase I after purification by column chromatography and electrophoretic analysis. Rat kidney cytosol was fractionated by ammonium sulfate precipitation, and the proteins were separated by ion-exchange column chromatography
Christoph M Ernst et al.
Molecular microbiology, 80(2), 290-299 (2011-02-11)
Bacteria are frequently exposed to cationic antimicrobial peptides (CAMPs) from eukaryotic hosts (host defence peptides) or from prokaryotic competitors (bacteriocins). However, many bacteria, among them most of the major human pathogens, achieve CAMP resistance by MprF, a unique enzyme that
Tao Dong et al.
Bioresource technology, 101(16), 6569-6571 (2010-04-07)
The effects of bovine serum albumin (BSA) addition on the cross-linked enzyme aggregates (CLEA) of aminoacylase from Aspergillus melleus (EC 3.5.1.14) were conducted at varying glutaraldehyde to enzyme ratio. After optimization, CLEA of aminoacylase prepared with 10 mg BSA per
Vildan Caner et al.
Antonie van Leeuwenhoek, 94(4), 527-532 (2008-07-31)
A total of 190 Campylobacter spp. isolates, of which 34 gave the result of very weak activity, and 156 gave the negative activity in the test for hippurate hydrolysis were characterized. The genomic DNA was isolated from a fresh culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service