Skip to Content
Merck
All Photos(1)

Key Documents

A263

Sigma-Aldrich

ATPA

solid

Synonym(s):

(RS)-2-Amino-3-(3-hydroxy-5-tert-butylisoxazol-4-yl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O4
CAS Number:
Molecular Weight:
228.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

DMSO: 10 mg/mL
H2O: 2.5 mg/mL

SMILES string

CC(C)(C)c1onc(O)c1CC(N)C(O)=O

InChI

1S/C10H16N2O4/c1-10(2,3)7-5(8(13)12-16-7)4-6(11)9(14)15/h6H,4,11H2,1-3H3,(H,12,13)(H,14,15)

InChI key

PIXJURSCCVBKRF-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Selective kainate receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Micale et al.
Journal of medicinal chemistry, 45(20), 4433-4442 (2002-09-20)
In this paper we describe the synthesis of a series of novel 2-[(4-alkylsemicarbazono)-(4-aminophenyl)-methyl]-4,5-methylenedioxyphenylacetic acid alkyl esters (10-19) carrying an alkylsemicarbazono moiety at a benzylic site. The influence of this group on the biological activity was evaluated by testing the corresponding
V R J Clarke et al.
Neuropharmacology, 42(7), 889-902 (2002-06-19)
Kainate receptors are involved in a variety of synaptic functions in the CNS including the regulation of excitatory synaptic transmission. Previously we described the depressant action of the GLU(K5) selective agonist (RS)-2-amino-3-(3-hydroxy-5-tert-butylisoxazol-4-yl)propanoic acid (ATPA) on synaptic transmission in the Schaffer
Marie-Louise Lunn et al.
Journal of medicinal chemistry, 46(5), 872-875 (2003-02-21)
Two X-ray structures of the GluR2 ligand-binding core in complex with (S)-2-amino-3-(5-tert-butyl-3-hydroxy-4-isoxazolyl)propionic acid ((S)-ATPA) have been determined with and without Zn(2+) ions. (S)-ATPA induces a domain closure of ca. 21 degrees compared to the apo form. The tert-butyl moiety of
Jon Valgeirsson et al.
Journal of medicinal chemistry, 46(26), 5834-5843 (2003-12-12)
A series of 2-arylureidobenzoic acids (AUBAs) was prepared by a short and effective synthesis, and the pharmacological activity at glutamate receptors was evaluated in vitro and in vivo. The compounds showed noncompetitive antagonistic activity at the kainate receptor subtype GluR5.
James S Robinson et al.
American journal of physiology. Heart and circulatory physiology, 282(6), H2371-H2376 (2002-05-11)
The hypothesis that glutamate dilates pial arterioles of newborn pigs through the production of carbon monoxide (CO) was addressed. Anesthesized newborn pigs were equipped with cranial windows to measure pial arteriolar responses to stimuli. Heme oxygenase (HO) inhibitors added topically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service