51269
Dihydroxyacetone phosphate hemimagnesium salt hydrate
≥95% (TLC)
Synonym(s):
1,3-Dihydroxy-2-propanone 1-phosphate hemimagnesium salt hydrate, 1-Hydroxy-3-(phosphonooxy)-2-propanone hemimagnesium salt hydrate, DHAP Mg
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H5Mg0.5O6P · xH2O
CAS Number:
Molecular Weight:
180.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥95% (TLC)
storage temp.
−20°C
SMILES string
O.OCC(=O)COP(O)(=O)O[Mg]OP(O)(=O)OCC(=O)CO
InChI
1S/2C3H7O6P.Mg.H2O/c2*4-1-3(5)2-9-10(6,7)8;;/h2*4H,1-2H2,(H2,6,7,8);;1H2/q;;+2;/p-2
InChI key
XIBMKSGIYJARCI-UHFFFAOYSA-L
Application
Dihydroxyacetone phosphate hemimagnesium salt hydrate (DHAP Mg) is a magnesium salt of DHAP. DHAP may be used as the substrate to characterize enzymes such as fructose bisphosphate aldolase and triose phosphate isomerase.
Biochem/physiol Actions
Dihydroxyacetone phosphate (DHAP) is a metabolic intermediate involved in many pathways, including glycolysis, gluconeogenesis, glycerol metabolism, phosphatidic acid synthesis, fat metabolism, and the Calvin cycle.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inés Ardao et al.
Langmuir : the ACS journal of surfaces and colloids, 28(15), 6461-6467 (2012-03-21)
Gold nanoparticles (AuNPs) are attractive materials for the immobilization of enzymes due to several advantages such as high enzyme loading, absence of internal diffusion limitations, and Brownian motion in solution, compared to the conventional immobilization onto porous macroscopic supports. The
Michael Fischer et al.
Chemical communications (Cambridge, England), 47(23), 6647-6649 (2011-05-13)
An efficient gram scale synthesis of 3-fluoro-1-hydroxyacetone phosphate (FHAP) has been developed. As a close analog to dihydroxyacetone phosphate, FHAP was used as a novel donor substrate for rabbit muscle aldolase catalyzed reactions. The different binding affinities of the gem-diol
Alice Chan et al.
Angewandte Chemie (International ed. in English), 51(31), 7711-7714 (2012-06-21)
Stop for NadA! A [4Fe-4S] enzyme, NadA, catalyzes the formation of quinolinic acid in de novo nicotinamide adenine dinucleotide (NAD) biosynthesis. A structural analogue of an intermediate, 4,5-dithiohydroxyphthalic acid (DTHPA), has an in vivo NAD biosynthesis inhibiting activity in E. coli. The
Matthias Bujara et al.
Nature chemical biology, 7(5), 271-277 (2011-03-23)
Recruiting complex metabolic reaction networks for chemical synthesis has attracted considerable attention but frequently requires optimization of network composition and dynamics to reach sufficient productivity. As a design framework to predict optimal levels for all enzymes in the network is
Bjørn O Hald et al.
The FEBS journal, 279(23), 4410-4420 (2012-10-18)
Coherent glycolytic oscillations in Saccharomyces cerevisiae are a multicellular property induced by addition of glucose to a starved cell population of sufficient density. However, initiation of oscillations requires an additional perturbation, usually addition of cyanide. The fate of cyanide during
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service