Skip to Content
Merck
All Photos(1)

Key Documents

50644

Sigma-Aldrich

N-Glycolylneuraminic acid

≥95% (HPLC)

Synonym(s):

Neu5Glc, NeuNGl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO10
CAS Number:
Molecular Weight:
325.27
Beilstein:
1716828
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[H][C@]1(O[C@@](O)(C[C@H](O)[C@H]1NC(=O)CO)C(O)=O)[C@H](O)[C@H](O)CO

InChI

1S/C11H19NO10/c13-2-5(16)8(18)9-7(12-6(17)3-14)4(15)1-11(21,22-9)10(19)20/h4-5,7-9,13-16,18,21H,1-3H2,(H,12,17)(H,19,20)/t4-,5+,7+,8+,9+,11-/m0/s1

InChI key

FDJKUWYYUZCUJX-AJKRCSPLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glycolylneuraminic acid, a component of non-human milk and colostrums, is used as a reference to analyse changes of the glycome during lactation. Glycolylneuraminic acid, a xenoantigen, is involved in antigenic reactions (antibody-mediated hyperacute rejection) during xenotransplantion.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Regulation of N-glycolylneuraminic acid biosynthesis in rat and mouse liver
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lepers et al.
European journal of biochemistry, 193(3), 715-723 (1990-11-13)
The relative contribution of N-glycoloyl-beta-D-neuraminic acid (Neu5Gc) to total sialic acids expressed in mouse and rat liver glycoconjugates was found to be 95% and 11%, respectively. This considerable difference in sialic acid composition made these two tissues suitable models for
Rachel E Taylor et al.
The Journal of experimental medicine, 207(8), 1637-1646 (2010-07-14)
The nonhuman sialic acid N-glycolylneuraminic acid (Neu5Gc) is metabolically incorporated into human tissues from certain mammalian-derived foods, and this occurs in the face of an anti-Neu5Gc "xeno-autoantibody" response. Given evidence that this process contributes to chronic inflammation in some diseases
Véronique Spichtig et al.
Analytical biochemistry, 405(1), 28-40 (2010-06-18)
Sialic acids are becoming recognized as important components of milk-based products for infants and young children. As such, many companies now label the sialic acid content of their products. To control the labeling, suitable methods are required for this analysis.
Qiaozhen Lu et al.
Analytical chemistry, 84(6), 2761-2768 (2012-03-07)
Human polyclonal IgG antibodies directly against the nonhuman sialic acid N-glycolylneuraminic acid (Neu5Gc) are potential biomarkers and mechanistic contributors to cancer and other diseases associated with chronic inflammation. Using a sialoglycan microarray, we screened the binding pattern of such antibodies
Potential impact of the non-human sialic acid N-glycolylneuraminic acid on transplant rejection risk.
Vered Padler-Karavani et al.
Xenotransplantation, 18(1), 1-5 (2011-02-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service