Skip to Content
Merck
All Photos(2)

Key Documents

504-05A

Sigma-Aldrich

Human Tracheal Epithelial Cells: HTEpC, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human trachea (normal surface epithelium)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

epithelial

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HTEpC provide a useful model system to study all aspects of epithelial function and disease, particularly those related to airway viral infections, as well as tissue repair mechanisms, signaling changes and potential treatments relevant to lung injuries, mechanical and oxidative stress, pulmonary diseases and smoking.

HTEpC have been used in a study investigating the role of goblet cell hyperplasia, possibly mediated by the expression of SPDEF, to the pathogenesis of chronic pulmonary diseases (Park, 2007).

Characterization: the cells have a characteristic morphology consistent with an epithelial origin and are positive for epithelial cell marker cytokeratin 18.

Cell Line Origin

Trachea

Application

epithelial function, tissue repair, cell signaling, lung injury treatment, mechanical and oxidative stress tests, lung lubrication, humidity maintenance, cleaning of respiratory tract, testing of drugs, toxins and carcinogens, respiratory function

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the HTEpC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service