Skip to Content
Merck
All Photos(1)

Key Documents

257M-1

Sigma-Aldrich

GCDFP-15 (23A3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

23A3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (257M-14)
vial of 0.5 mL concentrate (257M-15)
bottle of 1.0 mL predilute (257M-17)
vial of 1.0 mL concentrate (257M-16)
bottle of 7.0 mL predilute (257M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG2a

control

breast, breast carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PIP(5304)

General description

GCDFP-15 is a 15 kD glycoprotein which is localized in the apocrine metaplastic epithelium lining breast cysts and in apocrine glands in the axilla, vulva, eyelid, ear canal, and in salivary glands. Approximately 70% of breast carcinomas are labeled with antibody to GCDFP-15. Colorectal carcinomas, lung carcinoma, mesothelimas rarely stain with this antibody. Because of its specificity for breast carcinoma (up to 99%), this antibody is often used in a screening panel in the appropriate clinical situation, often for a woman with metastasis of unknown primary.

Quality


IVD

IVD

IVD

RUO

Linkage

GCDFP-15 Positive Control Slides, Product No. 257S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Viacava et al.
Virchows Archiv : an international journal of pathology, 432(3), 255-260 (1998-04-09)
GCDFP-15, a glycoprotein identified in the cyst fluid of cystic breast disease, is considered to be a marker of apocrine differentiation. Studies on GCDFP-15 localization in adult normal tissues are lacking, and no information on GCDFP-15 expression during fetal development
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 227-228 (2014)
A H Ormsby et al.
Journal of the American Academy of Dermatology, 32(5 Pt 1), 711-716 (1995-05-01)
For accurate classification of cutaneous metastatic carcinoma, reliable tumor-specific immunohistochemical markers would be valuable. Our purpose was to analyze the sensitivity, specificity, and positive predictive value of gross cystic disease fluid protein-15 (GCDFP-15) and estrogen receptor protein (ERP) for diagnosing
M R Wick et al.
Human pathology, 20(3), 281-287 (1989-03-01)
The identification of metastatic carcinoma of the breast may be difficult in the absence of a previous history of breast cancer. Various immunophenotypic markers have been introduced to aid in this process. A monoclonal antibody directed at a 15-kilodalton (kd)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service