Skip to Content
Merck
All Photos(1)

Key Documents

96466

Sigma-Aldrich

Zinc carbonate basic

purum p.a., ≥58% Zn basis (KT)

Synonym(s):

Zinc hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[ZnCO3]2 · [Zn(OH)2]3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

form

powder

concentration

≥58% Zn (KT)

solubility

water: insoluble

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

Ca: ≤5000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤200 mg/kg
K: ≤200 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg

SMILES string

[Zn++].[Zn++].O[Zn]O.O[Zn]O.O[Zn]O.[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/2CH2O3.6H2O.5Zn/c2*2-1(3)4;;;;;;;;;;;/h2*(H2,2,3,4);6*1H2;;;;;/q;;;;;;;;5*+2/p-10

InChI key

UOURRHZRLGCVDA-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Application

Zinc carbonate basic (basic zinc carbonate, [ZnCO3]2.[Zn(OH)2]3) may be used in the preparation of ZnO nanopowder. It may be employed as a catalyst for the synthesis of methyl phenylcarbamate, via reaction between aniline and dimethyl carbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thermal decomposition of basic zinc carbonate in nitrogen atmosphere.
Liu Y, et al.
Thermochimica Acta, 414(2), 121-123 (2004)
Highly selective phosgene-free carbamoylation of aniline by dimethyl carbonate under continuous-flow conditions.
Grego S, et al.
Organic Process Research & Development, 17(4), 679-683 (2013)
Synthesis of ZnO nanoparticles by thermal decomposition of basic zinc carbonate.
Hutera B, et al.
Archives of Metallurgy and Materials, 58(2), 489-491 (2013)
Cui Li et al.
Annals of botany, 123(1), 57-68 (2018-07-19)
The pathways whereby foliar-applied nutrients move across the leaf surface remain unclear. The aim of the present study was to examine the pathways by which foliar-applied Zn moves across the sunflower (Helianthus annuus) leaf surface, considering the potential importance of
Claudia Backes et al.
ACS nano, 13(6), 7050-7061 (2019-06-15)
Liquid phase exfoliation is a commonly used method to produce 2D nanosheets from a range of layered crystals. However, such nanosheets display broad size and thickness distributions and correlations between area and thickness, issues that limit nanosheet application potential. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service