Skip to Content
Merck
All Photos(1)

Key Documents

334340

Sigma-Aldrich

Molecular sieves, 13X

pellets, 1.6 mm diameter

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na86[AlO2)86(SiO2)106] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
SB.52

description

Length: approximately twice the particle diameter

Quality Level

form

pellets

concentration

13 × (miscellaneous)

diameter

1.6 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Molecular Sieve 13X is a highly porous sodium form of zeolite X suitable for the adsorption of large molecules such as aromatics, branched-chain hydrocarbons, and organosulfur compounds.

Application

Molecular Sieve 13X has been used to trap carbon dioxide for 14C (radiocarbon) analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of the adsorption of thiophenic sulfur compounds using flow calorimetry
Ng Flora TT, et al.
Applied Catalysis. B, Environmental, 56(1-2) (2005)
Advances in the radiocarbon analysis of carbon dioxide at the NERC radiocarbon facility (East Kilbride) using molecular sieve cartridges
Garnett MH, et al.
Radiocarbon, 61(6) (2019)
Adsorption and desorption experimental study of carbon dioxide/methane mixture gas on 13X-type molecular sieves
He Tianbiao, et al.
J. Chem. Eng. Jpn., 46(12), 811-820 (2013)
Purification and upgrading of biogas by pressure swing adsorption on synthetic and natural zeolites
Alonso-Vicario A, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 134(1-3), 100-107 (2010)
Adsorption of CO2 on molecular sieves and activated carbon.
Siriwardane RV, et al.
Energy and Fuels, 15(2), 279-284 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service