Skip to Content
Merck
All Photos(4)

Key Documents

274739

Sigma-Aldrich

Sand

50-70 mesh particle size

Synonym(s):

Sand, white quartz, Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

Agency

suitable for EPA 1613

form

granular

color

white

particle size

50-70 mesh

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sand, generally known as silicon dioxide, is used as a precursor, filler, and additive or processing aid in plastics, synthetic rubbers, catalyst carriers, chemical sensors, adhesives, pesticides, paper, paints, coatings, sealants, or insulation materials.

Application

Sand and granite can be employed as a catalyst in the three-component condensation of aldehydes, 1,3-dicarbonyl compounds, and ureas or thioureas to produce 3,4-dihydropyrimidinones and 3,4-dihydropyrimidinthiones.
Suitable for chromatography

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas hydrate formation in a variable volume bed of silica sand particles.
Linga P, et al.
Energy and Fuels, 23(11), 5496-5507 (2009)
B R Bickmore et al.
Environmental science & technology, 35(22), 4481-4486 (2002-01-05)
Caustic NaNO3 solutions containing dissolved Al were reacted with quartz sand at 89 degrees C to simulate possible reactions between leaked nuclear waste and primary subsurface minerals at the U.S. Department of Energy's Hanford site in Washington. Nitrate-cancrinite began to
Doris Dallinger et al.
Nature protocols, 2(2), 317-321 (2007-04-05)
We present here a protocol for the synthesis of the dihydropyrimidine (DHPM) derivative monastrol, which is known to be a specific mitotic kinesin Eg5 inhibitor. By applying controlled microwave heating under sealed-vessel conditions, the synthesis via the one-pot three-component Biginelli
Jenia Gutman et al.
Environmental science & technology, 47(1), 398-404 (2012-11-29)
This investigation is focused on the combined effect of bacterial physicochemical characteristics and motility on cell adhesion and deposition using a flow-through quartz crystal microbalance with dissipation (QCM-D). Three model flagellated strains with different degrees of motility were selected, including
B Afra et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(4), 045006-045006 (2012-12-15)
The morphology of swift heavy ion tracks in crystalline α-quartz was investigated using small angle x-ray scattering (SAXS), molecular dynamics (MD) simulations and transmission electron microscopy. Tracks were generated by irradiation with heavy ions with energies between 27 MeV and

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service