Skip to Content
Merck
All Photos(3)

Key Documents

244120

Sigma-Aldrich

Sodium perborate tetrahydrate

9-11% available Oxygen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBO3 · 4H2O
CAS Number:
Molecular Weight:
153.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder or granules

grade

for analytical purposes

Quality Level

form

powder or granules

quality

9-11% available Oxygen

concentration

~6.5-7.5% NaOH (titration)
~9-11% (active oxygen basis (by Na2S2O3, titration))

pH

10-10.4 (25 °C, 10 g/L)

mp

60 °C (dec.) (lit.)

SMILES string

O.O.O.O.[Na+].[O-]B1OO1

InChI

1S/BO3.Na.4H2O/c2-1-3-4-1;;;;;/h;;4*1H2/q-1;+1;;;;

InChI key

IBDSNZLUHYKHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium perborate is widely employed as an oxidizing agent.

Application

Sodium perborate tetrahydrate may be employed for the following studies:
  • Transformation of iodoarenes to (diacetoxyiodo)arenes.
  • Oxidation of Π-deficient azines to afford N-oxides.
  • Oxidation of sulphur containing heterocyclic compounds to afford S,S-dioxides.
  • Microwave-assisted oxidative hydration of various aromatic and aliphatic nitriles.
  • Various oxidation reactions of anilines and sulphides.
  • As a catalyst for the transformation of 2′-hydroxychalcones to flavanones.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium perborate tetrahydrate?mediated transformations of 2'-hydroxychalcones to flavanones, flavones, and 3', 5'-diiodoflavone under mild, environmentally friendly conditions.
Ganguly NC, et al.
Synthetic Communications, 43(10), 1351-1361 (2013)
Functional group oxidation using sodium perborate1.
McKillop A and Tarbin JA.
Tetrahedron, 43(8), 1753-1758 (1987)
Further functional group oxidations using sodium perborate.
McKillop A and Kemp D.
Tetrahedron, 45(11), 3299-3306 (1989)
Microwave-promoted transformation of nitriles to amides with aqueous sodium perborate.
Sharifi A, et al.
Synthetic Communications, 31(3), 431-434 (2001)
Viviane Maia Barreto de Oliveira et al.
Gerodontology, 28(2), 134-139 (2009-09-29)
To determine the effectiveness of a denture cleanser in reducing the concentration of volatile sulphur compounds (VSC) and its antimicrobial action. Micro-organisms from the denture biofilm can cause local and systemic disease and halitosis. Denture cleansers are important adjuncts in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service