Skip to Content
Merck
All Photos(1)

Key Documents

Y0001590

Metformin impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dicyandiamide, Cyanoguanidine, Dicyanodiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHCN
CAS Number:
Molecular Weight:
84.08
Beilstein:
605637
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metformin

manufacturer/tradename

EDQM

mp

208-211 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=N)NC#N

InChI

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

InChI key

QGBSISYHAICWAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metformin impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changkyu Kim et al.
Nanoscale, 7(15), 6627-6635 (2015-03-21)
The realization of air-stable nanoparticles, well-formulated nanoinks, and conductive patterns based on copper is a great challenge in low-cost and large-area flexible printed electronics. This work reports the synthesis of a conductively interconnected copper structure via thermal sintering of copper
Peng Li et al.
Angewandte Chemie (International ed. in English), 54(2), 574-577 (2014-11-15)
Self-assembly of a trigonal building subunit with diaminotriazines (DAT) functional groups leads to a unique rod-packing 3D microporous hydrogen-bonded organic framework (HOF-3). This material shows permanent porosity and demonstrates highly selective separation of C2H2/CO2 at ambient temperature and pressure.
Jon Draher et al.
Journal of chromatography. A, 1373, 106-113 (2014-12-02)
A method for the rapid quantification of 9 potential nitrogen-rich economic adulterants (dicyandiamide, urea, biuret, cyromazine, amidinourea, ammeline, amidinourea, melamine, and cyanuric acid) in five milk and soy derived nutritional ingredients, i.e. whole milk powder, nonfat dry milk, milk protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service