Skip to Content
Merck
All Photos(1)

Key Documents

Y0000011

Hymecromone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Methylumbelliferone, β-Methylumbelliferone, 4-MU, 7-Hydroxy-4-methylcoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Beilstein:
142217
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hymecromone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC(=O)Oc2cc(O)ccc12

InChI

1S/C10H8O3/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5,11H,1H3

InChI key

HSHNITRMYYLLCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hymecromone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etty Osher et al.
Orphanet journal of rare diseases, 10, 45-45 (2015-04-22)
Late Onset Tay- Sachs disease (LOTS) is a rare neurodegenerative lysosomal storage disease which results from mutations in the gene encoding the α subunit (HEXA) of β-hexosaminidase enzyme (HexA). At the present time, no effective treatment exists for LOTS and
Ching-Chieh Weng et al.
Molecular cancer, 18(1), 96-96 (2019-05-22)
The TG-interacting factor 1 (TGIF1) gene, which encodes a nuclear transcriptional corepressor of the TGFβ1/Smad signaling pathway, has been implicated in the pathogenesis of various types of human cancer; however, its role in pancreatic ductal adenocarcinoma (PDAC) has yet to
Melissa A Gray et al.
Nature chemical biology, 16(12), 1376-1384 (2020-08-19)
Currently approved immune checkpoint inhibitor therapies targeting the PD-1 and CTLA-4 receptor pathways are powerful treatment options for certain cancers; however, most patients across cancer types still fail to respond. Consequently, there is interest in discovering and blocking alternative pathways
Tae Kyung Hyun et al.
PloS one, 9(11), e112946-e112946 (2014-11-15)
Despite the completion of the Arabidopsis genome sequence, for only a relatively low percentage of the encoded proteins experimental evidence concerning their function is available. Plant proteins that harbour a single PLAT (Polycystin, Lipoxygenase, Alpha-toxin and Triacylglycerol lipase) domain and
Mi-Ok Sim et al.
Chemico-biological interactions, 216, 9-16 (2014-03-26)
This study was conducted to evaluate the effects of umbelliferone (UF) and 4-methylumbelliferone (mUF) on high-fat diet-induced hypertriglyceridemia and hyperglycemia in mice. The mice were assigned to normal control, high-fat control, and high-fat with UF or mUF groups. For UF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service