Skip to Content
Merck
All Photos(1)

Key Documents

W366412

Sigma-Aldrich

2,5-Dimethyl-4-methoxy-3(2H)-furanone

natural, ≥98%, FG

Synonym(s):

Mesifurane, strawberry furanone methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
FEMA Number:
3664
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.089
NACRES:
NA.21
Organoleptic:
caramel; jam; maple; musty; strawberry; sweet
grade:
FG
Halal
Kosher
natural
biological source:
sugar beets
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

sugar beets

Quality Level

grade

FG
Halal
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.478 (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; jam; maple; musty; strawberry; sweet

storage temp.

2-8°C

SMILES string

COC1=C(C)OC(C)C1=O

InChI

1S/C7H10O3/c1-4-6(8)7(9-3)5(2)10-4/h4H,1-3H3

InChI key

SIMKGHMLPVDSJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Classification of different pineapple varieties grown in Malaysia based on volatile fingerprinting and sensory analysis.: This research utilizes the 2,5-Dimethyl-4-methoxy-3(2H)-furanone compound in a study of pineapple varieties in Malaysia, analyzing volatile components to classify and differentiate varieties. The study contributes to agricultural biochemistry and plant science, enhancing understanding of plant varietal differences at the molecular level (Lasekan et al., 2018).

  • A glucosyltransferase specific for 4-hydroxy-2,5-dimethyl-3(2H)-furanone in strawberry.: This paper details the biochemical characterization of a glucosyltransferase enzyme in strawberries that specifically reacts with 2,5-Dimethyl-4-methoxy-3(2H)-furanone. Insights from this research are crucial for understanding enzymatic activity in fruit maturation and flavor development, providing valuable information for food science and agricultural chemistry (Yamada et al., 2018).

  • An oxidoreductase from ′Alphonso′ mango catalyzing biosynthesis of furaneol and reduction of reactive carbonyls.: This article explores an oxidoreductase enzyme in Alphonso mangoes that catalyzes reactions involving 2,5-Dimethyl-4-methoxy-3(2H)-furanone. The findings enhance our understanding of the biochemical pathways involved in aroma compound synthesis in mangoes, impacting agricultural biotechnology and flavor science (Kulkarni et al., 2013).

  • Natural 4-hydroxy-2,5-dimethyl-3(2H)-furanone (Furaneol).: This review provides an extensive overview of the naturally occurring 2,5-Dimethyl-4-methoxy-3(2H)-furanone, highlighting its significant role in flavor science. The study covers its biosynthesis, biological activities, and applications in enhancing the sensory properties of food products, making it highly relevant for researchers in the field of food chemistry and biochemistry (Schwab, 2013).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service