Skip to Content
Merck
All Photos(1)

Key Documents

T2200000

Trimethoprim

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Beilstein:
625127
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trimethoprim

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)

InChI key

IEDVJHCEMCRBQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christine Manyando et al.
PloS one, 9(5), e96017-e96017 (2014-05-17)
Antibiotic therapy during pregnancy may be beneficial and impacts positively on the reduction of adverse pregnancy outcomes. No studies have been done so far on the effects of daily Co-trimoxazole (CTX) prophylaxis on birth outcomes. A phase 3b randomized trial
Karin Tegmark Wisell et al.
The Journal of antimicrobial chemotherapy, 62(1), 35-40 (2008-04-15)
The lack of oral treatment alternatives for enterococcal urinary tract infections (UTIs) has led to a renewed interest in trimethoprim. Enterococci can incorporate exogenously produced folates and thereby reverse the effect of trimethoprim. Although a large proportion of enterococci appear
Sarah L Svensson et al.
PloS one, 9(8), e106063-e106063 (2014-08-29)
Campylobacter jejuni is a leading cause of foodbourne gastroenteritis, despite fragile behaviour under standard laboratory conditions. In the environment, C. jejuni may survive within biofilms, which can impart resident bacteria with enhanced stress tolerance compared to their planktonic counterparts. While
Luis H Braga et al.
The Journal of urology, 191(5 Suppl), 1501-1507 (2014-04-01)
We determined the feasibility of a clinical trial of continuous antibiotic prophylaxis to prevent urinary tract infections in children with prenatal hydronephrosis. We piloted a blinded, randomized, placebo controlled study of the urinary tract infection rate in infants with grades
Dolores Camacho-Muñoz et al.
Chemosphere, 111, 70-79 (2014-07-07)
The distribution of pharmaceutically active compounds in the environment has been reported in several works in which wastewater treatment plants have been identified as the main source of these compounds to the environment. The concentrations of these compounds in influent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service