Skip to Content
Merck
All Photos(3)

Key Documents

STS0204

Sigma-Aldrich

ECO TWEEN® 80

greener alternative

viscous liquid

Synonym(s):

TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.28
Pricing and availability is not currently available.

description

non-ionic

Quality Level

Assay

≥58%

form

viscous liquid

mol wt

micellar avg mol wt 79.000
average mol wt 1310

composition

Oleic acid, ≥58.0% (balance primarily linoleic, palmitic, and stearic acids)

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

0.012 mM (20-25°C)

viscosity

300—500 cP(25 °C)

transition temp

cloud point 65 °C

solubility

water: 100 mg/mL, clear to slightly hazy, faintly yellow

density

1.064 g/cm3

HLB

15.0

greener alternative category

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ECO Tween® 80, also known as Polyoxyethylenesorbitan monooleate, is a bio-based, nonionic detergent manufactured from a natural fatty acid. This surfactant, derived from natural sources, provides various functional advantages, such as detergency, emulsification, and wetting. Its versatility makes it well-suited for a variety of applications in biochemical and biological research, with common uses including cell lysis, protein extraction, nuclei isolation and cell fractionation.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.
TWEEN® 80 has been used in drug vehicle solution for specific injections in mice and rats.[1][2] It has been used for the determination of esterase activity in bacterial cells.[3]

Biochem/physiol Actions

TWEEN® 80 is a polysorbate surfactant which helps in stabilizing proteins and in protein membrane studies. It is regularly used for the extraction of membrane proteins and isolation of nuclei from cells.[4][5][6]

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ECO TWEEN is a registered trademark of Croda Inc.
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HU-210 shows higher efficacy and potency than morphine after intrathecal administration in the mouse formalin test.
Guhring H et al.
European Journal of Pharmacology, 429, 127-127 (2001)
Pathogenic factors of Pseudomonas cepacia isolates from patients with cystic fibrosis.
Gessner AR and Mortensen JE
Journal of Medical Microbiology, 33, 115-115 (1990)
Scopes RK
Protein Purification: Principles and Practice (1993)
V Ryu et al.
Neuroscience, 167(4), 1227-1238 (2010-03-04)
Previous reports show that vagal afferent innervation of the stomach eventually regenerates from surviving nodose ganglion (NG) neurons after subdiaphragmatic vagotomy. Systemic capsaicin treatment destroys gastric vagal afferent neurons expressing vanilloid receptor 1 (VR1). However, it is not known whether
Handbook of Biochemistry and Molecular Biology (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service