Skip to Content
Merck
All Photos(1)

Key Documents

PHR3473

Supelco

Azacitidine Related Compound A

certified reference material, pharmaceutical secondary standard

Synonym(s):

5-Azacytosine, 2-Hydroxy-4-aminotriazine, 4-Amino-1,3,5-triazin-2-ol, 4-Amino-1,3,5-triazin-2-one, 4-Amino-2-hydroxy-1,3,5-triazine, NSC 51100, NSC 54006

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N4O
CAS Number:
Molecular Weight:
112.09
Beilstein:
116378
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1045539

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

mp

>300 °C (lit.)

application(s)

pharmaceutical

SMILES string

NC1=NC(=O)NC=N1

InChI

1S/C3H4N4O/c4-2-5-1-6-3(8)7-2/h1H,(H3,4,5,6,7,8)

InChI key

MFEFTTYGMZOIKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

Reactant for:
  • Enzymic synthesis of nucleoside analogs using immobilized 2′-deoxyribosyltransferase from Lactobacillus reuteri
  • Reactions with oxide radical ion

  • Potential antitumor and antiproliferative agent against human leukemia cells

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service