Skip to Content
Merck
All Photos(1)

Key Documents

PHR1732

Supelco

Vancomycin hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Vancomycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. V0045000
traceable to USP 1709007

API family

vancomycin

CofA

current certificate can be downloaded

packaging

pkg of 4x250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42+,44-,46+,47+,48-,49+,50-,51+,52+,53+,54-,56+,57+,65-,66-;/m0./s1

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Vancomycin hydrochloride is a glycopeptide antibiotic, effective for the treatment of endocarditis caused by Gram-positive bacteria. It is used to inhibit the synthesis of bacterial cell wall and prevents the synthesis of peptidoglycan by binding tightly to peptides containing D-alanyl-D-alanine at the free carboxyl end. It is isolated from both Streotomyces orientalis and Nocardia lurida.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Vancomycin hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations, pure form and injections preparations using high-performance liquid chromatography technique and spectrophotometric technique.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0718 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow Injection-Spectrophotometric Determination of Vancomycin Hydrochloride in Pharmaceutical Preparations Using Diazotized Metoclopramide
Al-Abachi QM, et al.
Journal of Al-Nahrain University, 18(1), 9-19 (2015)
James Shaw et al.
Journal of orthopaedic trauma, 34(4), e109-e113 (2020-03-21)
Prefabrication and storage of antibiotic beads may decrease surgical time and allow for use in other settings. This study investigated the effects of sterilization technique and storage time on the bioactivity of antibiotic polymethyl methacrylate (PMMA) beads. Uniform beads of
Development and validation of high-performance liquid chromatographic method for the simultaneous determination of ceftriaxone and vancomycin in pharmaceutical formulations and biological samples
Tariq A, et al.
ScienceAsia, 36(4), 297-304 (2010)
Andrés Aranda-Díaz et al.
eLife, 9 (2020-01-30)
Predicting antibiotic efficacy within microbial communities remains highly challenging. Interspecies interactions can impact antibiotic activity through many mechanisms, including alterations to bacterial physiology. Here, we studied synthetic communities constructed from the core members of the fruit fly gut microbiota. Co-culturing
Lina Fan et al.
Gut microbes, 13(1), 1-17 (2021-02-10)
Inflammatory bowel disease (IBD) is defined as an immune dysregulation disease with poor prognosis. Various therapies based on gut microbe modulation have been proposed. In this study, we aim to explore the therapeutic effect of B. adolescentis on IBD, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service