Skip to Content
Merck
All Photos(1)

Key Documents

PHR1210

Supelco

Cumene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(1-Methylethyl)benzene, 2-Phenylpropane, Isopropylbenzene, NSC 8776

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)2
CAS Number:
Molecular Weight:
120.19
Beilstein:
1236613
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1151709

vapor density

4.1 (vs air)

vapor pressure

8 mmHg ( 20 °C)
9.7 mmHg ( 37.7 °C)

API family

cumene

CofA

current certificate can be downloaded

autoignition temp.

797 °F

expl. lim.

6.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.491 (lit.)

bp

152-154 °C (lit.)

mp

−96 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)c1ccccc1

InChI

1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

RWGFKTVRMDUZSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Cumene may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9023 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

A novel peroxy radical based oxidative stressing system for ranking the oxidizability of drug substances
Harmon PA. et al.
Journal of Pharmaceutical Sciences, 95(9), 2014-2028 (2006)
Determination of Residual Solvents in Bulk Drug and Formulations
Singh P and Sinha M
American Journal of PharmTech Research, 3(4), 289-295 (2006)
Johanna Bråred Christensson et al.
Contact dermatitis, 55(4), 230-237 (2006-09-09)
Concomitant positive reactions to colophonium, oxidized limonene, and/or oxidized linalool are recorded in patch test studies. The main allergens in these patch test mixtures are hydroperoxides, which form antigens by a radical pathway. Theoretically, concomitant reactions can be explained not
Mathieu Frenette et al.
Journal of the American Chemical Society, 128(51), 16432-16433 (2006-12-21)
A new class of thermally activated chain-breaking antioxidants is presented. Dimers of persistent carbon-centered radicals are able to inhibit the autoxidation of cumene and styrene with better rate constants than the commercial antioxidant Irganox HP-136 and 3,5-di-tert-butyl-4-hydroxyanisole. A dramatic increase
C Holvoet et al.
Die Pharmazie, 62(2), 126-132 (2007-03-08)
Paclitaxel, an antitumoral drug, is poorly soluble in aqueous media. Therefore, in a commercialised formulation (Taxol), paclitaxel (30 mg active compound) is dissolved in polyethoxylated castor oil (Cremophor EL) and ethanol. After dilution of Taxol in aqueous media paclitaxel tends

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service