Skip to Content
Merck
All Photos(1)

Key Documents

P3850000

Pseudoephedrine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-Pseudoephedrine hydrochloride, (+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
3915112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pseudoephedrine

manufacturer/tradename

EDQM

mp

185-188 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10+;/m0./s1

InChI key

BALXUFOVQVENIU-KXNXZCPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pseudoephedrine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Minjie Jiang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 974, 126-130 (2014-12-03)
A rapid, sensitive and selective high-performance liquid chromatography-tandem mass spectrometric method (HPLC-MS) has been developed and validated for the simultaneous determination of 14-thienyl methylene matrine (TMM) and matrine (MT) in rat plasma in the present study. The analytes were separated
Minjie Jiang et al.
PloS one, 10(2), e0116010-e0116010 (2015-02-26)
A rapid, sensitive and selective high-performance liquid chromatography-tandem mass spectrometric method (HPLC-MS) was developed and validated to determine the 14-(3-methylbenzyl)matrine (3MBM) and 14-(4-methylbenzyl)matrine (4MBM) levels in rat plasma in the present study. The analytes were separated using a C18 column

Articles

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service