Skip to Content
Merck
All Photos(1)

Key Documents

NIST1570A

Trace elements in spinach leaves

NIST® SRM® 1570a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1570a contains 60g of powdered dried spinach leaves.

SRM 1570A_cert

SRM 1570A_SDS

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction values and coverage factor (k) for the elements in SRM1570a.
  • The non-certified and information values of elements are reported as mass fraction.
  • The certified values are traceable to the International System of Units (SI) derived unit of mass fraction, expressed as percent.
  • The NIST certificate is provided with expiration certificate, use, storage, safety, and maintenance instructions.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Nitrogen (N), Phosphorous
Trace Elements: Aluminum (Al), Arsenic (As), Boron (B), Cadmium (Cd), Calcium (Ca), Cobalt (Co), Copper (Cu), Europium (Eu), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Nickel (Ni), Potassium (K), Rubidium (Rb), Scandium (Sc), Selenium (Se), Sodium (Na), Strontium (Sr), Sulfur (S), Thorium (Th), Uranium (U), Vanadium (V), Zinc (Zn)

Matrix Group: Fruits and Vegetables

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service