Skip to Content
Merck
All Photos(2)

Key Documents

L3900

Sigma-Aldrich

2,6-Lutidine

ReagentPlus®, 98%

Synonym(s):

2,6-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
105690
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

98%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

143-145 °C ((lit.))
143-145 °C (lit.)

mp

-6 °C
−6 °C (lit.)

density

0.92  g/mL at 25 °C ((lit.))
0.92 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)ON1C(=O)CCC1=O
Cc1cccc(C)n1

InChI

1S/C16H27NO4/c1-2-3-4-5-6-7-8-9-10-11-16(20)21-17-14(18)12-13-15(17)19/h2-13H2,1H3
1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3

InChI key

LRKGVVJOUNKTGG-UHFFFAOYSA-N


OISVCGZHLKNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Lutidine can be used as a base for the oxidation of α-CF3 alcohols to the corresponding carbonyl compounds in the presence of 4-acetamido-2,2,6,6-tetramethylpiperidine-1-oxoammonium tetrafluoroborate (Bobbitt′s salt) as oxidation reagent.
It can be used:
  • As a promoter for catalytic asymmetric fluorination of α-cyanophosphonates in the presence of chiral Pd(II)-bisphosphine complexes.
  • In combination with tert-butyldimethylsilyl triflate for the protection of tertiary alcohols and unreactive secondary alcohols.
  • In combination with triethylsilyl trifluoromethanesulfonate for the conversion of acetals to the corresponding aldehydes in dichloromethane followed by workup in water.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unexpected Highly Chemoselective Deprotection of the Acetals from Aldehydes and Not Ketones: TESOTf? 2, 6-Lutidine Combination.
Fujioka H, et al.
Journal of the American Chemical Society, 126(38), 11800-11801 (2004)
Pd (II)-Catalyzed Asymmetric Fluorination of ?-Aryl-?-cyanophosphonates with the Aid of 2, 6-Lutidine.
Moriya K I, et al.
Synlett, 2007(07), 1139-1142 (2007)
Studies with trialkylsilyltriflates: new syntheses and applications.
Corey E J, et al.
Tetrahedron Letters, 22(36), 3455-3458 (1981)
Synthesis of 4-acetamido-2, 2, 6, 6-tetramethylpiperidine-1-oxoammonium tetrafluoroborate and 4-acetamido-(2, 2, 6, 6-tetramethyl-piperidin-1-yl) oxyl and their use in oxidative reactions.
Mercadante M A, et al.
Nature Protocols, 8(4), 666-666 (2013)
Bojana Ginovska et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(44), 15713-15719 (2015-10-24)
We report that 2,6-lutidine⋅trichloroborane (Lut⋅BCl3 ) reacts with H2 in toluene, bromobenzene, dichloromethane, and Lut solvents producing the neutral hydride, Lut⋅BHCl2 . The mechanism was modeled with density functional theory, and energies of stationary states were calculated at the G3(MP2)B3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service