Skip to Content
Merck
All Photos(1)

Key Documents

L0505000

Levomepromazine maleate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28N2O5S
CAS Number:
Molecular Weight:
444.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levomepromazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S1c2c(cc(cc2)OC)N(c3c1cccc3)C[C@@H](CN(C)C)C.OC(=O)\C=C/C(=O)O

InChI

1S/C19H24N2OS.C4H4O4/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;5-3(6)1-2-4(7)8/h5-11,14H,12-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-;/m1./s1

InChI key

IFLZPECPTYCEBR-VIEYUMQNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levomepromazine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Kumazawa et al.
Journal of chromatography. A, 1218(18), 2521-2527 (2011-03-25)
Solid-phase extraction (SPE) using micropipette tips is a useful technique to prepare samples prior to mass spectrometry. However, most commercial SPE tips have loading capacities that are insufficient for quantitative determination. In this paper, we describe a rapid method for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service