Skip to Content
Merck
All Photos(1)

Key Documents

G4028

Supelco

Ginkgolide A from Ginkgo biloba leaves

analytical standard, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O9
CAS Number:
Molecular Weight:
408.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥90% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[H][C@]12CC34[C@@]5([H])C[C@@H](C(C)(C)C)C36[C@@H](O)C(=O)O[C@@]6([H])OC4(C(=O)O5)[C@@]1(O)[C@H](C)C(=O)O2

InChI

1S/C20H24O9/c1-7-12(22)26-10-6-17-9-5-8(16(2,3)4)18(17)11(21)13(23)28-15(18)29-20(17,14(24)27-9)19(7,10)25/h7-11,15,21,25H,5-6H2,1-4H3/t7-,8+,9-,10+,11+,15+,17?,18?,19-,20?/m1/s1

InChI key

FPUXKXIZEIDQKW-ZJZLDMJOSA-N

General description

Ginkgolide A is a diterpene lactone, which can be extracted from the leaves of Ginkgo biloba tree.

Application

Ginkgolide A may be used as a standard in the quantitative determination of ginkgolide A, extracted from Ginkgo biloba leaves using reverse phase high performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Constituent of Ginkgo biloba extracts. Antioxidant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Supercritical fluids extraction of Ginkgo ginkgolides and flavonoids
Chiu L K, et al.
Journal of Supercritical Fluids, 24(1), 77-87 (2002)
Progress in Drug Research 56 (2012)
Determination of ginkgolides and bilobalide in Ginkgo biloba leaves and phytopharmaceuticals
Beek V A T, et al.
Journal of Chromatography A, 543, 375-387 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service