Skip to Content
Merck
All Photos(1)

Key Documents

G1522

Sigma-Aldrich

Gentamicin

solution, suitable for hybridoma, BioReagent

Synonym(s):

Garamycin, Gentiomycin C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
42020823
PubChem Substance ID:
NACRES:
NA.76

product name

Gentamicin solution, Hybri-Max, 50 mg/mL gentamicin, 0.1 μm filtered, BioReagent, suitable for hybridoma

grade

Hybri-Max

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

quality

Prepared in tissue culture grade water.

concentration

50 mg/mL gentamicin

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

color

colorless to light yellow

density

1.000 g/cm3

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@H](OC[C@]1(C)O)O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](CN)CC[C@H]3N)[C@@H]2O.CN[C@@H]4[C@@H](O)[C@H](OC[C@]4(C)O)O[C@H]5[C@H](N)C[C@H](N)[C@@H](O[C@H]6O[C@@H](CC[C@H]6N)[C@@H](C)N)[C@@H]5O.CN[C@H](C)[C@@H]7CC[C@@H](N)[C@H](O7)O[C@@H]8[C@@H](N)C[C@@H](N)[C@H](O[C@H]9OC[C@](C)(O)[C@H](NC)[C@H]9O)[C@H]8O

InChI

1S/C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;1-8(21)12-5-4-9(22)18(30-12)31-15-10(23)6-11(24)16(13(15)26)32-19-14(27)17(25-3)20(2,28)7-29-19;1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;1-5(2,3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;8-19,25-28H,4-7,21-24H2,1-3H3;8-18,24-27H,3-7,20-23H2,1-2H3;(H2,1,2,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;8-,9-,10+,11-,12+,13+,14-,15-,16+,17-,18-,19-,20+;8-,9+,10-,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m110./s1

InChI key

RDEIXVOBVLKYNT-HDZPSJEVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Gentamicin solution has been used to study its dose-related antipruritic effects on serotonin (5-HT)-induced scratching model. It has also been used in electrophysiological analysis.

Biochem/physiol Actions

Mode of Action: Gentamicin inhibits protein synthesis by binding to the 30S subunit of the ribosome.

Antimicrobial spectrum: Includes Gram-negative and Gram-positive bacteria, including strains resistant to tetracycline, chloramphenicol, kanamycin and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus and Streptococcus.

Components

Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt. There are three components, each consisting of five basic nitrogens and requiresing five equivalents of sulfuric acid per mole of gentamicin base.

Caution

The product is stable at 37° for 5 days and is stored at 2-8°C.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-term depression induced by endogenous cannabinoids produces neuroprotection via astroglial CB1R after stroke in rodents
<BIG>Wang F, et al.</BIG>
Journal of Cerebral Blood Flow and Metabolism, 1122-1137 (2019)
Meng Wei et al.
Biochemical and biophysical research communications, 503(2), 1154-1159 (2018-06-29)
Like tumour cells, during intraerythrocytic stage, Plasmodium-infected erythrocytes rely completely on glucose absorption from the blood circulation for energy metabolism. Glucose is taken up by glucose transporter 1 (GLUT1) on human red blood cells (RBCs) and glucose transporter 4 (GLUT4)
Laurent Vidard et al.
Journal of immunology (Baltimore, Md. : 1950), 203(3), 676-685 (2019-06-16)
To understand and dissect the mechanisms driving human NK cell proliferation, we exploited the methodology used in cell therapy to numerically expand NK cells in the presence of K562-derived artificial APC (aAPCs) and cytokines. For four consecutive weeks, high expression
Mulualem E Tilahun et al.
PloS one, 6(11), e27203-e27203 (2011-11-22)
Staphylococcal enterotoxin B (SEB) is one of a family of toxins secreted by Staphylococcus aureus that act as superantigens, activating a large fraction of the T-cell population and inducing production of high levels of inflammatory cytokines that can cause toxic
Lingyan Wang et al.
Molecular vision, 17, 2835-2846 (2011-12-01)
There is an unmet challenge to promote wound healing in non-healing wounds such as in the post-LASIK (laser-assisted in situ keratomileusis) cornea. Using human corneal fibroblasts (HCFs) in cell culture, we investigated the concentration dependence of the growth factor transforming

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service