Skip to Content
Merck
All Photos(1)

Key Documents

C1700000

Chlorothiazide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClN3O4S2
CAS Number:
Molecular Weight:
295.72
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorothiazide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=S(C1=C2C=C(Cl)C(S(N)(=O)=O)=C1)(NC=N2)=O

InChI

1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)

InChI key

JBMKAUGHUNFTOL-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

One of the first in the thiazide class of diuretics.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte K Leech et al.
Acta crystallographica. Section B, Structural science, 64(Pt 1), 101-107 (2008-01-22)
The compounds chlorothiazide and hydrochlorothiazide (crystalline form II) have been studied in their fully hydrogenous forms by powder neutron diffraction on the GEM diffractometer. The results of joint Rietveld refinement of the structures against multi-bank neutron and single-bank X-ray powder
Krzysztof J Paluch et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 41(5), 603-611 (2010-09-08)
Chlorothiazide (CTZ), unlike other thiazide diuretics, can form salts. An injectable formulation containing the sodium salt is available; however neither the physicochemical characteristics of the salt nor its solid state form have been previously reported. This work reports on the
L P Brion et al.
The Cochrane database of systematic reviews, (1)(1), CD001454-CD001454 (2008-02-07)
Lung edema may complicate respiratory distress syndrome (RDS) in preterm infants. The aim of this review was to assess the risks and benefits of diuretic administration in preterm infants with RDS. The standard search method of the Cochrane Neonatal Review
Krzysztof J Paluch et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 42(3), 220-229 (2010-12-01)
Chlorothiazide (CTZ) is a poorly soluble diuretic agent. The aim of the present work was to produce and characterise a potassium salt form of chlorothiazide which has the potential advantages of improved aqueous solubility and potassium supplementation. A number of
Khalid Hussain et al.
The Journal of clinical endocrinology and metabolism, 91(8), 2839-2842 (2006-05-11)
Congenital hyperinsulinism (CHI) is a cause of severe hypoglycemia in the neonatal and infancy period. Histologically, there are two subtypes with diffuse and focal disease. The preoperative differentiation of these two forms is very important because the surgical management is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service