Skip to Content
Merck
All Photos(1)

Key Documents

BCR306

1-Nitronaphthalene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
1867714
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

304 °C (lit.)

mp

53-57 °C (lit.)

density

1.223 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cccc2ccccc12

InChI

1S/C10H7NO2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

RJKGJBPXVHTNJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Nitronaphthalene, belonging to the class of nitrated-polycyclic aromatic hydrocarbons, is persistent in the environment. It is produced from direct sources such as diesel, gasoline exhaust and gas-phase reactions of polycyclic aromatic hydrocarbons (PAHs) with oxides of nitrogen.

Analysis Note

For more information please see:
BCR306

Legal Information

BCR is a registered trademark of European Commission

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination and comparison of nitrated-polycyclic aromatic hydrocarbons measured in air and diesel particulate reference materials
Bamford AH, et al.
Chemosphere, 50 (5), 575-587 (2003)
Myong Gyong Lee et al.
American journal of respiratory cell and molecular biology, 38(3), 300-309 (2007-09-29)
1-Nitronaphthalene (1-NN) and ozone are cytotoxic air pollutants commonly found as components of photochemical smog. The mechanism of toxicity for 1-NN involves bioactivation by cytochrome P450s and subsequent adduction to proteins. Previous studies have shown that 1-NN toxicity in the
J Azmi et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 10(6), 401-416 (2005-11-26)
Metabolic fingerprints, in the form of patterns of high-concentration endogenous metabolites, of 1-nitronaphthalene (NN)-induced lung toxicity have been elucidated in bronchoalveolar lavage fluid (BALF), urine, blood plasma, and intact lung and liver tissue using NMR spectroscopy-based metabolic profiling. A single
Kara R Schmelzer et al.
Environmental health perspectives, 114(9), 1354-1360 (2006-09-13)
Ambient air is polluted with a mixture of pulmonary toxicants. Previous studies indicate that prior exposure to atmospheric oxidant pollutants such as ozone may significantly alter the response to other pollutants, such as 1-nitronaphthalene (1-NN) . 1-NN, a component of
Ching Yu Lin et al.
Proteomics, 6(3), 972-982 (2006-02-03)
Naphthalene and 1-nitronaphthalene are ambient air pollutants, which undergo P450-dependent bioactivation in the lung. Reactive metabolites of naphthalene and 1-nitronaphthalene covalently bind to proteins, and the formation of covalent adducts correlates with airway epithelial cell injury in rodent models. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service