Skip to Content
Merck
All Photos(1)

Key Documents

B8806

Sigma-Aldrich

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester

1 mg/mL in DMSO

Synonym(s):

BCECF-AM solution, Spiro(isobenzofuran-1(3H),9′-(9H)xanthene)-2′,7′-dipropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

description

Mixture of six isomeric compounds

Quality Level

form

liquid

concentration

1 mg/mL in DMSO

color

colorless to yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CCc1cc2c(Oc3cc(OCOC(C)=O)c(CCC(=O)OCOC(C)=O)cc3C24OC(=O)c5cc(ccc45)C(=O)OCOC(C)=O)cc1OCOC(C)=O

InChI

1S/C42H40O21/c1-22(43)52-17-57-34-15-36-32(13-27(34)7-10-38(48)59-19-54-24(3)45)42(31-9-6-29(12-30(31)41(51)63-42)40(50)61-21-56-26(5)47)33-14-28(8-11-39(49)60-20-55-25(4)46)35(16-37(33)62-36)58-18-53-23(2)44/h6,9,12-16H,7-8,10-11,17-21H2,1-5H3

InChI key

NTECHUXHORNEGZ-UHFFFAOYSA-N

Application

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester has been used for the estimation of vacuolar pH and intracellular pH.

Biochem/physiol Actions

An intracellular fluorescent pH indicator.
Hydrolyzed to BCECF by cytosolic esterases.

Physical form

Solution in 0.1% BCECF AM Ester and 99.9% DMSO

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Salil Chanroj et al.
The Journal of biological chemistry, 286(39), 33931-33941 (2011-07-29)
The complexity of intracellular compartments in eukaryotic cells evolved to provide distinct environments to regulate processes necessary for cell proliferation and survival. A large family of predicted cation/proton exchangers (CHX), represented by 28 genes in Arabidopsis thaliana, are associated with
A K Ho et al.
The American journal of physiology, 261(4 Pt 1), C642-C649 (1991-10-01)
In rat pinealocytes, activation of alpha 1-adrenergic receptors leads to increases in intracellular pH (pHi). In this study, the role of pHi on adrenergic regulation of cyclic nucleotide accumulation was investigated using ammonium chloride, which increased pHi, and sodium propionate
Viraj P Nirwan et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-03-07)
Herein, we report the fabrication and characterization of novel polycaprolactone (PCL)-based nanofibers functionalized with bare (ligand-free) titanium nitride (TiN) nanoparticles (NPs) for tissue engineering applications. Nanofibers were prepared by a newly developed protocol based on the electrospinning of PCL solutions
Chengzhi Hu et al.
Lab on a chip, 17(4), 671-680 (2017-01-18)
Pollen tubes are tip-growing plant cells that deliver the sperm cells to the ovules for double fertilization of the egg cell and the endosperm. Various directional cues can trigger the reorientation of pollen tube growth direction on their passage through
Evaluation of intracellular pH regulation and alkalosis defense mechanisms in preimplantation embryos.
Dagilgan S, et al.
Theriogenology, 83, 1075-1084 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service