Skip to Content
Merck
All Photos(1)

Key Documents

94119

Supelco

Glycine

analytical standard, for nitrogen determination according to Kjeldahl method

Synonym(s):

2-Aminoacetic acid, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0%

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
titration: suitable

impurities

18.6%±0.2% total nitrogen acc. to Kjeldahl

loss

≤0.05% loss on drying

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

1 M HCl: 75 mg/L, clear, colorless (2.252g in 30mL)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

NH4+: ≤200 mg/kg

application(s)

food and beverages

format

neat

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The glycine standard is produced under stringent conditions and measured with the highest possible precision.

Application

It can be used as an analytical standard to:
  • Evaluate Kjeldahl′s procedure to determine total protein content of reference standards used in clinical laboratories.
  • Analyze beer and wine for their protein and total usable nitrogen using a microwell ninhydrin assay.
  • Compare Kjeldahl and Dumas methods for routine analysis of dairy products in an interlaboratory study.
  • Simultaneously determine total phosphorus and total Kjeldahl nitrogen in surface water samples.
  • Quantitatively determine nitrate in surface waters using Valdetrama method.
  • Measure low level Kjeldahl nitrogen using an AutoAnalyzer.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Features and Benefits

  • Available in a neat format in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • High-quality offering accurate titer determinations

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of Glycine Dihyrate: Its Implications to Crystallization of Glycine from Solution and Modification of Glycine in Space
Hu.T.C, et al.
Acta Crystallographica Section A, Foundations of Crystallography, 73 (2017)
The Structure of Glycine Dihydrate: Implications for the Crystallization of Glycine from Solution and Its Structure in Outer Space
Xu W, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 129, 2062-2066 (2017)
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Li-Chun Tu et al.
The EMBO journal, 32(24), 3220-3230 (2013-11-12)
Nuclear pore complexes (NPCs) mediate cargo traffic between the nucleus and the cytoplasm of eukaryotic cells. Nuclear transport receptors (NTRs) carry cargos through NPCs by transiently binding to phenylalanine-glycine (FG) repeats on intrinsically disordered polypeptides decorating the NPCs. Major impediments
Suzanne L Grant et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 844(2), 278-282 (2006-08-08)
A simple and versatile methodology using high-performance liquid chromatography (HPLC) with fluorimetric detection was developed to simultaneously determine d-serine along with other metabolically related neuroactive amino acids in the glutamatergic system: L-serine, L-glutamate, L-glutamine, and glycine. On-column sensitivity was in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service