90161
(2,2′)-Furildioxime
for spectrophotometric det. of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII), U(VI), ≥97.0% (sum of isomers, HPLC)
Synonym(s):
Bis(2-furyl)glyoxime
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8N2O4
CAS Number:
Molecular Weight:
220.18
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
description
mixture of isomers
Assay
≥97.0% (sum of isomers, HPLC)
form
crystals
technique(s)
UV/Vis spectroscopy: suitable
SMILES string
O\N=C(c1ccco1)\C(=N\O)c2ccco2
InChI
1S/C10H8N2O4/c13-11-9(7-3-1-5-15-7)10(12-14)8-4-2-6-16-8/h1-6,13-14H/b11-9+,12-10+
InChI key
RBOZTFPIXJBLPK-WGDLNXRISA-N
General description
Oximes are selective reagents which react with limited number of metal ions. These form water-insoluble complexes extractable into organic solvents. Furildioxime, a gravimetric and colorimetric reagent for nickel, palladium, platinum and rhenium is prepared by the oximation of furil with hydroxyammonium chloride followed by aqueous recrystallisation using MacKair′s method.
Application
a-furildioxime [(2,2′)-Furildioxime] was used:
- as neutral carrier in Calcium(II)-selective potentiometric sensor, selective electrode used for titration.
- in the synthesis of ketenimines and bis(ketenimines).
- in determination of palladium in water samples by rapid spectrophotometric analysis.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Calcium (II)-selective potentiometric sensor based on a-furildioxime as neutral carrier.
Singh, Ashok Kumar, and Sameena Mehtab.
Sensors and Actuators B, Chemical, 123, 429-436 (2007)
D L Bissett et al.
Journal of the American Academy of Dermatology, 31(4), 572-578 (1994-10-01)
In previous work we found that iron is a factor in skin photodamage, apparently by way of its participation in oxygen radical production. Here we report topical photoprotection with the iron chelator 2-furildioxime (FDO). Our purpose was to determine the
The furildioximes. Part I. The structure of the isomeric furildioximes.
Fryer, F. A., D. J. B. Galliford, and J. T. Yardley.
Analyst, 88, 188-190 (1963)
Single-Channel Flow Injection Spectrophotometric Determination of Nickel Using Furildioxime in Micellar Solution.
Memon, Najma, et al.
TheScientificWorldJournal, 12, 1-5 (2012)
Three-component reaction of alkyl isocyanides with acetylenic esters and pyridine-2-carboxaldoxime or a-furildioxime: Synthesis and dynamic NMR study of ketenimines and bis (ketenimines).
Baharfar, Robabeh, Leyla Jaafari, and Razieh Azimi.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 22, 943-946 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service