Skip to Content
Merck
All Photos(1)

Key Documents

86859

Sigma-Aldrich

Tetrabutylammonium hydroxide 30-hydrate

≥99.0% (T)

Synonym(s):

N,N,N-Tributyl-1-butanaminium hydroxide triacontahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH) · 30H2O
CAS Number:
Molecular Weight:
799.93
Beilstein:
3571228
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (T)

form

crystals

impurities

≤0.05% halides (as bromide)

mp

27-30 °C (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.31H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h5-16H2,1-4H3;31*1H2/q+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-1

InChI key

DFGIRVKFXSRUOX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium hydroxide 30-hydrate is a strong alkali containing quaternary amine, which is used in a variety of organic transformations.

Application

Tetrabutylammonium hydroxide 30-hydrate can be used:
  • As a base for the chemoselective allenylation of aldehydes with TMS-protected alkynes to synthesize α-allenols.
  • As an additive for the hydroxyalkylation of phenols with cyclic carbonates to synthesize aryl β-hydroxyethyl ethers.
  • In the solvothermal synthesis of SNS (simple nanoscroll structures) and INS(intercalated nanoscrolls) hexaniobate nanoscrolls.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid solvothermal fabrication of hexaniobate nanoscrolls.
Adireddy S, et al.
Materials Research Bulletin, 48(9), 3236-3241 (2013)
Alana K Greaves et al.
Chemosphere, 93(3), 574-580 (2013-08-08)
Perfluorooctane sulfonate (PFOS) is a globally distributed persistent organic pollutant that has been found to bioaccumulate and biomagnify in aquatic food webs. Although principally in its linear isomeric configuration, 21-35% of the PFOS manufactured via electrochemical fluorination is produced as
Nils Erik Halvorsen et al.
Pharmaceuticals (Basel, Switzerland), 13(2) (2020-02-15)
A simple color spot test for determining the presence of residual tetrabutylammonium (TBA) in various 18F-radiopharmaceuticals is described. The test can be performed in less than five minutes. Iodoplatinate-saturated TLC plates are initially spotted with the 18F-radiopharmaceutical to be tested
Carmen Cavallo et al.
Scientific reports, 5, 16785-16785 (2015-11-19)
Solid solutions of the rare earth (RE) cations Pr(3+), Nd(3+), Sm(3+), Gd(3+), Er(3+) and Yb(3+) in anatase TiO2 have been synthesized as mesoporous beads in the concentration range 0.1-0.3% of metal atoms. The solid solutions were have been characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service