Skip to Content
Merck
All Photos(2)

Key Documents

78364

Supelco

Brilliant Green

analytical standard

Synonym(s):

Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
Colour Index Number:
42040
Beilstein:
3901207
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

210 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OS([O-])(=O)=O.CCN(CC)c1ccc(cc1)C(\c2ccccc2)=C3/C=CC(\C=C3)=[N+](/CC)CC

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant green is a triphenylmethane dye, found to be toxic to humans and animals. It is widely used in dying wool, leather, silk, jute, and cotton. It is also used in the manufacturing of green ink, as a staining constituent of bacteriological media.

Application

Brilliant green may be used as a reference standard for the determination of the analyte in grass carp tissues using enzyme linked immunosorbent assay technique (ELISA).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applicability of waste materials-bottom ash and deoiled soya-as adsorbents for the removal and recovery of a hazardous dye, brilliant green.
Mittal A, et al.
Journal of Colloid and Interface Science, 326(1), 8-17 (2008)
Yu-Dong Shen et al.
Analytica chimica acta, 707(1-2), 148-154 (2011-10-27)
An immunizing hapten (4-(carboxymethoxy)phenyl)bis(4-(diethylamino)phenyl)methylium for brilliant green (BG), a triphenylmethane dye with a potential illegal use in fish feeding, was synthesized and used to produce polyclonal antibody (PcAb) against BG. Unexpectedly, the obtained PcAb showed high cross-reactivity (CR) to malachite
R Salsoso et al.
Placenta, 36(3), 287-296 (2015-01-13)
Preeclampsia is associated with impaired placental vasodilation and reduced endothelial nitric oxide synthase (eNOS) activity in the foetoplacental circulation. Adenosine and insulin stimulate vasodilation in endothelial cells, and this activity is mediated by adenosine receptor activation in uncomplicated pregnancies; however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service