Skip to Content
Merck
All Photos(3)

Key Documents

414255

Sigma-Aldrich

1-Chlorobutane

anhydrous, 99.5%

Synonym(s):

Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Cl
CAS Number:
Molecular Weight:
92.57
Beilstein:
1730909
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.5%
bp:
77-78 °C (lit.)
vapor pressure:
80.1 mmHg ( 78.4 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.2 (vs air)

vapor pressure

80.1 mmHg ( 78.4 °C)

Assay

99.5%

form

liquid

autoignition temp.

860 °F

expl. lim.

10.1 %

impurities

<0.002% water

evapn. residue

<0.0003%

refractive index

n20/D 1.402 (lit.)

bp

77-78 °C (lit.)

mp

−123 °C (lit.)

density

0.886 g/mL at 25 °C (lit.)

SMILES string

CCCCCl

InChI

1S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3

InChI key

VFWCMGCRMGJXDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chlorobutane is an isomer of chlorobutane.[1] It undergoes stoichiometric and catalytic dehydrochlorination on reacting with nanocrystalline MgO (magnesium oxide) to generate isomers of butene.[2] Viscosities of the binary mixtures of pentyl acetate and 1-chlorobutane and 1-chlorobutane and acetonitrile has been studied. This study helps in better understanding of cosolvency in polymer system.[3]

Application

1-Chlorobutane may be used in the synthesis of 1-butyl-3-methylimidazolium hydrogen sulfate ([BMIM][HSO4]).[4]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous Prediction of Densities and Vapor-Liquid Equilibria of Mixtures Containing an Isomeric Chlorobutane and Methyl tert-Butyl Ether Using the VTPR Model.
Montano D, et al.
Industrial & Engineering Chemistry Research, 50(24), 14193-14202 (2011)
Viscosities and excess volumes of binary mixtures formed by the liquids acetonitrile, pentyl acetate, 1-chlorobutane, and carbon tetrachloride at 25?C.
Prolongo MG, et al.
The Journal of Physical Chemistry, 88(10), 2163-2167 (1984)
Extraction and oxidative desulfurization of diesel fuel catalyzed by a Br?nsted acidic ionic liquid at room temperature.
Gao H, et al.
Green Chemistry, 12(7), 1220-1224 (2010)
Changes in texture and catalytic activity of nanocrystalline MgO during its transformation to MgCl2 in the reaction with 1-chlorobutane.
Fenelonov VB, et al.
The Journal of Physical Chemistry B, 105(18), 3937-3941 (2001)
M E Alburges et al.
Journal of analytical toxicology, 19(6), 381-386 (1995-10-01)
Ibogaine, an indolamine derivative, is currently being investigated as a potential agent in the treatment of stimulant and opiate addiction. We developed a rapid, sensitive, and specific method for the analysis of ibogaine and its putative active metabolite, 12-hydroxy-ibogamine (12-OH-ibogamine).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service