Skip to Content
Merck
All Photos(1)

Key Documents

39436

Supelco

Arsenic Standard for AAS

TraceCERT®, 1 g/L As in nitric acid (nominal concentration), Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

1 g/L As in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Monocrotaline-induced Vasomotor Dysfunction: A study demonstrated that pulmonary hypertension impairs vasomotor function in rat diaphragm arterioles, with monocrotaline being used to induce the condition, thereby aiding in understanding the systemic vascular effects of pulmonary arterial hypertension (Schulze et al., 2024).


Analysis Note

1 g/L As in 2% nitric acid (contains NaOH), prepared with high purity As2O3, HNO3, NaOH and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zygmunt M Gusiatin et al.
Journal of hazardous materials, 338, 160-166 (2017-06-02)
Batch washing experiments were performed to evaluate the feasibility of using a solution of humic substances (HS) extracted from municipal sewage sludge as a washing agent to remove As from soils at a former As mining area. Soils (S1, S2

Articles

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service